Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-2-Piperidinecarboxylic acid, 97+%, Acros Organics

Click to view available options
Quantity:
1g
5g
50g
Packaging:
Glass bottle
98303-20-9
C11H19NO4
229.28
MFCD01862877
JQAOHGMPAAWWQO-UHFFFAOYSA-N
n-boc-2-piperidinecarboxylic acid,1-tert-butoxycarbonyl piperidine-2-carboxylic acid,1-boc-piperidine-2-carboxylic acid,n-boc-dl-pipecolinic acid,n-boc-piperidine-2-carboxylic acid,boc-dl-pip-oh,n-boc-pipecolic acid,1-n-boc-2-piperidinecarboxylic acid,1-tert-butoxy carbonyl piperidine-2-carboxylic acid,1-tert-butoxycarbonyl-2-piperidinecarboxylic acid
581831
1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CCCCC1C(=O)O
98303-20-9
C11H19NO4
229.28
MFCD01862877
JQAOHGMPAAWWQO-UHFFFAOYSA-N
n-boc-2-piperidinecarboxylic acid,1-tert-butoxycarbonyl piperidine-2-carboxylic acid,1-boc-piperidine-2-carboxylic acid,n-boc-dl-pipecolinic acid,n-boc-piperidine-2-carboxylic acid,boc-dl-pip-oh,n-boc-pipecolic acid,1-n-boc-2-piperidinecarboxylic acid,1-tert-butoxy carbonyl piperidine-2-carboxylic acid,1-tert-butoxycarbonyl-2-piperidinecarboxylic acid
581831
1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CCCCC1C(=O)O
Melting Point 132°C to 134°C
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Formula Weight 229.28
Physical Form Crystalline Powder
Percent Purity 97+%
Chemical Name or Material N-BOC-2-Piperidinecarboxylic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning