Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-Piperidine-4-carbonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
91419-52-2
C11H18N2O2
210.28
UQADQTBQNVARAP-UHFFFAOYSA-N
1-boc-4-cyanopiperidine,1-n-boc-4-cyanopiperidine,n-boc-4-cyanopiperidine,1-n-boc-4-cyano-piperidine,n-boc-4-cyano-piperidine,1-piperidinecarboxylic acid, 4-cyano-, 1,1-dimethylethyl ester,boc-4-cyanopiperidine,4-cyano-piperidine-1-carboxylic acid tert-butyl ester,tert-butyl 4-cyano-1-piperidinecarboxylate,n-boc-piperidine-4-carbonitrile
1514443
tert-butyl 4-cyanopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C#N
91419-52-2
C11H18N2O2
210.28
UQADQTBQNVARAP-UHFFFAOYSA-N
1-boc-4-cyanopiperidine,1-n-boc-4-cyanopiperidine,n-boc-4-cyanopiperidine,1-n-boc-4-cyano-piperidine,n-boc-4-cyano-piperidine,1-piperidinecarboxylic acid, 4-cyano-, 1,1-dimethylethyl ester,boc-4-cyanopiperidine,4-cyano-piperidine-1-carboxylic acid tert-butyl ester,tert-butyl 4-cyano-1-piperidinecarboxylate,n-boc-piperidine-4-carbonitrile
1514443
tert-butyl 4-cyanopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C#N
Packaging Glass Bottle
Melting Point 59°C to 63°C
Infrared Spectrum Authentic
Color White
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 210.28
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material N-BOC-Piperidine-4-carbonitrile
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning