Learn More
N-Boc-propargylamine, 97%, Thermo Scientific Chemicals
Description
N-Boc-propargylamine is used to prepare triazolobenzylidene-thiazolopyrimidines. which act as CDC25 phosphatase inhibitors. Further, it is used for the synthesis of beta-glucan polysaccharide analogs. In addition to this, it is involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-disubstituted cyclopentenones .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Boc-propargylamine is used to prepare triazolobenzylidene-thiazolopyrimidines. which act as CDC25 phosphatase inhibitors. Further, it is used for the synthesis of beta-glucan polysaccharide analogs. In addition to this, it is involved in the Pauson-Khand (PK) reaction of norbornadiene and N-Boc-propargylamine to prepare 4,5-disubstituted cyclopentenones .
Solubility
Soluble in chloroform.
Notes
Moisture sensitive. Incompatible with oxidizing agents. Store in a cool place.
Specifications
Melting Point | 40°C to 44°C |
Linear Formula | HC≡CCH2NHCO2C(CH3)3 |
Solubility Information | Soluble in chloroform. |
Formula Weight | 155.2 |
Percent Purity | 97% |
Quantity | 5 g |
Flash Point | 93°C (199°F) |
Sensitivity | Moisture sensitive |
Chemical Name or Material | N-Boc-propargylamine |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.