Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-Pyrrole, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
5176-27-2
C9H13NO2
167.21
IZPYBIJFRFWRPR-UHFFFAOYSA-N
tert-butyl 1h-pyrrole-1-carboxylate,n-boc-pyrrole,n-t-boc-pyrrole,tert-butyl 1-pyrrolecarboxylate,t-butyl 1h-pyrrole-1-carboxylate,1h-pyrrole-1-carboxylic acid, 1,1-dimethylethyl ester,n-tert-butoxycarbonyl-1h-pyrrole,1-boc pyrrole,1-boc-pyrrole,acmc-209kvp
643494
tert-butyl pyrrole-1-carboxylate
CC(C)(C)OC(=O)N1C=CC=C1
5176-27-2
C9H13NO2
167.21
IZPYBIJFRFWRPR-UHFFFAOYSA-N
tert-butyl 1h-pyrrole-1-carboxylate,n-boc-pyrrole,n-t-boc-pyrrole,tert-butyl 1-pyrrolecarboxylate,t-butyl 1h-pyrrole-1-carboxylate,1h-pyrrole-1-carboxylic acid, 1,1-dimethylethyl ester,n-tert-butoxycarbonyl-1h-pyrrole,1-boc pyrrole,1-boc-pyrrole,acmc-209kvp
643494
tert-butyl pyrrole-1-carboxylate
CC(C)(C)OC(=O)N1C=CC=C1
Specific Gravity 1
Color Colorless to Yellow
Density 1.0000g/mL
Quantity 100g
Flash Point 75°C
Boiling Point 97.0°C (2.3 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Formula Weight 167.21
Physical Form Solution
Percent Purity 98%
Chemical Name or Material N-BOC-Pyrrole
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning