Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-S-(3-nitro-2-pyridylthio)-L-cysteine, 95%

Click to view available options
Quantity:
1g
76880-29-0
C13H17N3O6S2
375.414
MFCD00153301
OVTLOLNDKQUMRH-QMMMGPOBSA-N
boc-cys npys-oh,t-butyloxycarbonyl-s-3-nitro-2-pyridinesulfenyl cysteine,t-bcnp,l-alanine,n-1,1-dimethylethoxy carbonyl-3-3-nitro-2-pyridinyl dithio,boc-cys npys,2r-2-tert-butoxycarbonyl amino-3-3-nitropyridin-2-yl disulfanyl propanoic acid,bcnp,n-tert-butoxycarbonyl-3-3-nitropyridin-2-yl disulfanyl-l-alanine,boc-cys npys-oh tlc,n,a-boc-s-3-nitro-2-pyridylthio-l-cysteine
131299
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[(3-nitropyridin-2-yl)disulfanyl]propanoic acid
CC(C)(C)OC(=O)NC(CSSC1=C(C=CC=N1)[N+](=O)[O-])C(=O)O
76880-29-0
C13H17N3O6S2
375.414
MFCD00153301
OVTLOLNDKQUMRH-QMMMGPOBSA-N
boc-cys npys-oh,t-butyloxycarbonyl-s-3-nitro-2-pyridinesulfenyl cysteine,t-bcnp,l-alanine,n-1,1-dimethylethoxy carbonyl-3-3-nitro-2-pyridinyl dithio,boc-cys npys,2r-2-tert-butoxycarbonyl amino-3-3-nitropyridin-2-yl disulfanyl propanoic acid,bcnp,n-tert-butoxycarbonyl-3-3-nitropyridin-2-yl disulfanyl-l-alanine,boc-cys npys-oh tlc,n,a-boc-s-3-nitro-2-pyridylthio-l-cysteine
131299
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[(3-nitropyridin-2-yl)disulfanyl]propanoic acid
CC(C)(C)OC(=O)NC(CSSC1=C(C=CC=N1)[N+](=O)[O-])C(=O)O
Applications
N-Boc-S-(3-nitro-2-pyridylthio)-L-cysteine is used as medical intermediates.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture. Protect from humidity and water. Incompatible with oxidizing agents.
Beilstein 3569570
Formula Weight 375.41
Physical Form Powder
Percent Purity 95%
Color Yellow
Melting Point ~160°C (decomposition)
Quantity 1g
Sensitivity Moisture Sensitive
Chemical Name or Material N-Boc-S-(3-nitro-2-pyridylthio)-L-cysteine