Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-sarcosine methyl ester, 97%

Click to view available options
Quantity:
250mg
1g
5g
42492-57-9
C9H17NO4
203.238
MFCD04973115
LTBDTYJYKRLTRN-UHFFFAOYSA-N
methyl 2-tert-butoxycarbonyl methyl amino acetate,n-boc-sarcosine methyl ester,boc-gly-ome,boc-sarcosine methyl ester,ltbdtyjykrltrn-uhfffaoysa,n-t-butoxycarbonyl-sarcosine methyl ester,n-tert-butoxycarbonylsarcosine methyl ester,methyl n-tert-butoxycarbonyl-n-methylglycinate,methyl 2-tert-butoxy carbonyl methyl amino acetate
14018930
methyl 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetate
CC(C)(C)OC(=O)N(C)CC(=O)OC
42492-57-9
C9H17NO4
203.238
MFCD04973115
LTBDTYJYKRLTRN-UHFFFAOYSA-N
methyl 2-tert-butoxycarbonyl methyl amino acetate,n-boc-sarcosine methyl ester,boc-gly-ome,boc-sarcosine methyl ester,ltbdtyjykrltrn-uhfffaoysa,n-t-butoxycarbonyl-sarcosine methyl ester,n-tert-butoxycarbonylsarcosine methyl ester,methyl n-tert-butoxycarbonyl-n-methylglycinate,methyl 2-tert-butoxy carbonyl methyl amino acetate
14018930
methyl 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetate
CC(C)(C)OC(=O)N(C)CC(=O)OC
Applications
N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.

Notes
Incompatible with oxidizing agents. Heat sensitive.
Linear Formula (CH3)3COCON(CH3)CH2CO2CH3
Formula Weight 203.24
Physical Form Liquid
Percent Purity 97%
Color Yellow
Quantity 1g
Chemical Name or Material N-Boc-sarcosine methyl ester

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold