Alfa Aesar™ N-Boc-trans-4-fluoro-L-proline methyl ester, 97%

Manufacturer: Alfa Aesar™ H2774703

Quantity 1g

 View more versions of this product

Catalog No. AAH2774703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 203866-18-6
    Molecular Formula C11H18FNO4
    Formula Weight 247.27
    MDL Number MFCD07367993
    Synonym (2S,4R)-4-Fluoropyrrolidine-1,2-dicarboxylic acid 1-tert-butyl 2-methyl diester
    Chemical Name or Material N-Boc-trans-4-fluoro-L-proline methyl ester

    Specifications

    Formula Weight 247.27
    Molecular Formula C11H18FNO4
    Melting Point 36° to 39°C
    MDL Number MFCD07367993
    CAS 203866-18-6
    Quantity 1g
    Synonym (2S,4R)-4-Fluoropyrrolidine-1,2-dicarboxylic acid 1-tert-butyl 2-methyl diester
    Recommended Storage 2° to 8°C
    Chemical Name or Material N-Boc-trans-4-fluoro-L-proline methyl ester
    Assay 97%

    Description

    Applications
    N-Boc-trans-4-fluoro-L-proline methyl ester is used to prepare (S)-2-[N-(N ft.-benzyl-(S)-4-fluoro-L-prolyl)amino]benzophenone by reacting with of 2-aminobenzophenone in the presence of potassium tert -butoxide.

    Solubility
    Insoluble in water.

    Notes
    Incompatible with strong oxidizing agents and strong bases.

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit