Alfa Aesar™ N-Boc-trans-4-hydroxy-D-proline, 99%

Manufacturer: Alfa Aesar™ H27155MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27155MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 147266-92-0
    Molecular Formula C10H17NO5
    Formula Weight 231.25
    MDL Number MFCD01861341
    Synonym Boc-D-Hyp-OH; 1-tert-Butoxycarbonyl-(2R,4S)-4-hydroxypyrrolidine-2-carboxylic acid
    Chemical Name or Material N-Boc-trans-4-hydroxy-D-proline

    Specifications

    Formula Weight 231.25
    Molecular Formula C10H17NO5
    Melting Point ≈123°C
    MDL Number MFCD01861341
    Quantity 250mg
    Synonym Boc-D-Hyp-OH; 1-tert-Butoxycarbonyl-(2R,4S)-4-hydroxypyrrolidine-2-carboxylic acid
    Recommended Storage 2° to 8°C
    CAS 147266-92-0
    Chemical Name or Material N-Boc-trans-4-hydroxy-D-proline
    Assay 99%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit