Alfa Aesar™ N-Boc-trans-4-hydroxy-L-proline, 97%

Manufacturer: Alfa Aesar™ H2711003

Quantity 1g

 View more versions of this product

Catalog No. AAH2711003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13726-69-7
    Molecular Formula C10H17NO5
    Formula Weight 231.25
    MDL Number MFCD00053370
    Synonym Boc-Hyp-OH; 1-tert-Butoxycarbonyl-(2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
    Chemical Name or Material N-Boc-trans-4-hydroxy-L-proline

    Specifications

    Formula Weight 231.25
    Molecular Formula C10H17NO5
    Melting Point 123° to 127°C
    CAS 13726-69-7
    Quantity 1g
    MDL Number MFCD00053370
    Synonym Boc-Hyp-OH; 1-tert-Butoxycarbonyl-(2S,4R)-4-hydroxypyrrolidine-2-carboxylic acid
    Chemical Name or Material N-Boc-trans-4-hydroxy-L-proline
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit