Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-trans-4-Hydroxy-L-proline methyl ester, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
74844-91-0
C11H19NO5
245.275
MFCD00076981
MZMNEDXVUJLQAF-SFYZADRCSA-N
boc-hyp-ome,n-boc-trans-4-hydroxy-l-proline methyl ester,2s,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate,1-tert-butyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate,boc-l-hydroxyproline methyl ester,boc-hydroxyproline-ome,methyl trans-1-boc-4-hydroxy-l-prolinate,n-tert-butoxycarbonyl-trans-4-hydroxy-l-proline methyl ester,trans-1-tert-butoxycarbonyl-4-hydroxy-l-proline methyl ester,n-boc-trans-4-hydroxyl-l-proline methyl ester
2734883
1-O-tert-butyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O
74844-91-0
C11H19NO5
245.275
MFCD00076981
MZMNEDXVUJLQAF-SFYZADRCSA-N
boc-hyp-ome,n-boc-trans-4-hydroxy-l-proline methyl ester,2s,4r-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate,1-tert-butyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate,boc-l-hydroxyproline methyl ester,boc-hydroxyproline-ome,methyl trans-1-boc-4-hydroxy-l-prolinate,n-tert-butoxycarbonyl-trans-4-hydroxy-l-proline methyl ester,trans-1-tert-butoxycarbonyl-4-hydroxy-l-proline methyl ester,n-boc-trans-4-hydroxyl-l-proline methyl ester
2734883
1-O-tert-butyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O
Quantity 1g
Packaging Glass bottle
Specific Rotation Condition −65° (20°C c=1 Chloroform)
Specific Rotation -63° to -67° (20°C, 589nm) (c=1, CHCl3)
Color White to Beige
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 245.28
Physical Form Crystalline Powder
Percent Purity ≥96%
Chemical Name or Material N-BOC-trans-4-Hydroxy-L-proline methyl ester
Show More Show Less