Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Bromoacetamide, 95%

Click to view available options
Quantity:
5g
25g
79-15-2
C2H4BrNO
137.964
MFCD00037097
VBTQNRFWXBXZQR-UHFFFAOYSA-N
acetamide, n-bromo,unii-0aq6mwh7zb,ccris 4590,0aq6mwh7zb,acetobromamide,n-bromo-acetamide,sjyhiabiktp@,acmc-209pgg,n-bromoacetamide, powder,lopac-b-2377
4353
N-bromoacetamide
CC(=O)NBr
79-15-2
C2H4BrNO
137.964
MFCD00037097
VBTQNRFWXBXZQR-UHFFFAOYSA-N
acetamide, n-bromo,unii-0aq6mwh7zb,ccris 4590,0aq6mwh7zb,acetobromamide,n-bromo-acetamide,sjyhiabiktp@,acmc-209pgg,n-bromoacetamide, powder,lopac-b-2377
4353
N-bromoacetamide
CC(=O)NBr
Applications
Abolishes the rapid inactivation of membrane sodium- and potassium-ion channels.

Solubility
Soluble in water.

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Melting Point 102°C to 106°C
Quantity 5g
Linear Formula CH3CONHBr
Beilstein 969346
Merck Index 14,1398
Solubility Information Soluble in water.
Formula Weight 137.97
Percent Purity 95%
Chemical Name or Material N-Bromoacetamide

GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 201-181-0

TSCA : Yes

Recommended Storage : Keep cold