Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Bromophthalimide 95%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
2439-85-2
C8H4BrNO2
226.029
MFCD00005888
MARXMDRWROUXMD-UHFFFAOYSA-N
n-bromophthalimide,2-bromoisoindoline-1,3-dione,2-bromo-1h-isoindole-1,3 2h-dione,1h-isoindole-1,3 2h-dione, 2-bromo,n-bromophtalimide,n-bromo phthalimide,acmc-1cjlt,2-bromobenzo c azoline-1,3-dione,1h-isoindole-1,3 2h-dione,2-bromo
75542
2-bromoisoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)Br
2439-85-2
C8H4BrNO2
226.029
MFCD00005888
MARXMDRWROUXMD-UHFFFAOYSA-N
n-bromophthalimide,2-bromoisoindoline-1,3-dione,2-bromo-1h-isoindole-1,3 2h-dione,1h-isoindole-1,3 2h-dione, 2-bromo,n-bromophtalimide,n-bromo phthalimide,acmc-1cjlt,2-bromobenzo c azoline-1,3-dione,1h-isoindole-1,3 2h-dione,2-bromo
75542
2-bromoisoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)Br
C-X bond formation reagent
Color Yellow
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 226.03
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material N-Bromophthalimide
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.