Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Bromosuccinimide, 99%, Thermo Scientific Chemicals

Catalog Number AAA1592230
Click to view available options
Quantity:
250 g
1000 g
5000 g
128-08-5
C4H4BrNO2
177.985
MFCD00005510
PCLIMKBDDGJMGD-UHFFFAOYSA-N
n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide
67184
CHEBI:53174
1-bromopyrrolidine-2,5-dione
C1CC(=O)N(C1=O)Br
128-08-5
C4H4BrNO2
177.985
MFCD00005510
PCLIMKBDDGJMGD-UHFFFAOYSA-N
n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide
67184
CHEBI:53174
1-bromopyrrolidine-2,5-dione
C1CC(=O)N(C1=O)Br

N-Bromosuccinimide is a chemical reagent used in electrophilic additions and radical substitution reactions in synthetic organic chemistry. In Wohl-Ziegler reaction, it is involved in allylic and benzylic bromination reaction. It is also used in the alfa-bromination of carbonyl derivatives. In the Hofmann rearrangement, it is used in the presence of a strong base, such as 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) reacts with primary amides to produce a carbamate. It oxidizes silyl ethers to aldehydes in the presence of azobisisobutyronitrile. It is also used for the modification of ribosomal sulfhydryl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Bromosuccinimide is a chemical reagent used in electrophilic additions and radical substitution reactions in synthetic organic chemistry. In Wohl-Ziegler reaction, it is involved in allylic and benzylic bromination reaction. It is also used in the alfa-bromination of carbonyl derivatives. In the Hofmann rearrangement, it is used in the presence of a strong base, such as 1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) reacts with primary amides to produce a carbamate. It oxidizes silyl ethers to aldehydes in the presence of azobisisobutyronitrile. It is also used for the modification of ribosomal sulfhydryl groups.

Solubility
Soluble in acetone, tetrahydrofuran, dimethyl formamide, dimethyl sulfoxide and acetonitrile. Slightly soluble in water and acetic acid. Insoluble in ether, hexane and carbon tetrachloride.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, iron, iron salts, halogenated hydrocarbons, amines and metals. Store in a cool place.
TRUSTED_SUSTAINABILITY
Melting Point 175°C to 180°C (decomposition)
Density 2.098
Assay Percent Range 99%
Quantity 250 g
UN Number UN3261
Beilstein 113916
Sensitivity Light sensitive
Merck Index 14,1438
Solubility Information Soluble in acetone,tetrahydrofuran,dimethyl formamide,dimethyl sulfoxide and acetonitrile. Slightly soluble in water and acetic acid. Insoluble in ether,hexane and carbon tetrachloride.
Formula Weight 177.99
Percent Purity 99%
Chemical Name or Material N-Bromosuccinimide
Show More Show Less
Hazard Category H272-H290-H315-H317-H319
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P220-P221-P234-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 204-877-2
TSCA Yes
storageNote1 Moisture sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit