Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Butoxymethyl)acrylamide, AcroSeal™, 97%, Thermo Scientific™

Catalog Number ac435641000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal glass Bottle
1852-16-0
C8H15NO2
157.213
UTSYWKJYFPPRAP-UHFFFAOYSA-N
n-butoxymethyl acrylamide, 2-propenamide, n-butoxymethyl, n-butoxymethylacrylamide, n-butoxymethylakrylamid, n-butoxymethyl-2-propenamide, acrylamide, n-butoxymethyl, n-n-butoxymethyl acrylamide, unii-z89g4o958a, n-butoxymethyl prop-2-enamide, n-butoxymethylakrylamid czech
15817
N-(butoxymethyl)prop-2-enamide
CCCCOCNC(=O)C=C
1852-16-0
C8H15NO2
157.213
UTSYWKJYFPPRAP-UHFFFAOYSA-N
n-butoxymethyl acrylamide, 2-propenamide, n-butoxymethyl, n-butoxymethylacrylamide, n-butoxymethylakrylamid, n-butoxymethyl-2-propenamide, acrylamide, n-butoxymethyl, n-n-butoxymethyl acrylamide, unii-z89g4o958a, n-butoxymethyl prop-2-enamide, n-butoxymethylakrylamid czech
15817
N-(butoxymethyl)prop-2-enamide
CCCCOCNC(=O)C=C

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point -9.0°C to +6.0°C
Color Colorless
Density 0.9600g/mL
Boiling Point 121.0°C to 124.0°C (0.1 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging AcroSeal glass Bottle
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
Quantity 100 mL
Specific Gravity 0.96
Stabilizer stabilised with MEHQ
Formula Weight 157.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-(Butoxymethyl)acrylamide
Show More Show Less
Hazard Statement GHS H Statement:
May cause cancer.
May cause genetic defects.
Suspected of damaging the unborn child.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Obtain special instructions before use.
IF exposed or concerned: Get medical attention/advice.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit