Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Butyl butyrate, 99%

Manufacturer:  Alfa Aesar™ B24390AK

Quantity 250mL

 View more versions of this product

Catalog No. AAB24390AK

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
It is very important for the food and beverages industries and used as perfuming agents. It is a applied as a solvent for resins.

Solubility
Soluble in water. (1 g/L).

Notes
Store at room temperature. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

109-21-7
CH3CH2CH2CO2(CH2)3CH3
MFCD00009450
1747101
Butyric acid n-butyl ester
C8H16O2
144.21
UN3272
14,1556
n-Butyl butyrate

Specifications

109-21-7
CH3CH2CH2CO2(CH2)3CH3
−92°C
UN3272
1747101
49°C (120°F)
Butyric acid n-butyl ester
99%
C8H16O2
144.21
MFCD00009450
164° to 165°C
250mL
14,1556
n-Butyl butyrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

RTECSNumber : ES8120000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit