Alfa Aesar™ n-Butyl butyrate, 99%

Manufacturer: Alfa Aesar™ B24390AU

Quantity 1000mL

 View more versions of this product

Catalog No. AAB24390AU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-21-7
    Molecular Formula C8H16O2
    Linear Formula CH3CH2CH2CO2(CH2)3CH3
    Formula Weight 144.21
    MDL Number MFCD00009450
    UN Number UN3272
    Beilstein 1747101
    Merck Index 14,1556
    Synonym Butyric acid n-butyl ester
    Chemical Name or Material n-Butyl butyrate

    Specifications

    Formula Weight 144.21
    Quantity 1000mL
    Flash Point 49°C (120°F)
    TSCA TSCA
    Molecular Formula C8H16O2
    Linear Formula CH3CH2CH2CO2(CH2)3CH3
    Melting Point −92°C
    MDL Number MFCD00009450
    UN Number UN3272
    Boiling Point 164° to 165°C
    Beilstein 1747101
    Merck Index 14,1556
    Synonym Butyric acid n-butyl ester
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number ES8120000
    CAS 109-21-7
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Chemical Name or Material n-Butyl butyrate
    Assay 99%

    Description

    Applications
    It is very important for the food and beverages industries and used as perfuming agents. It is a applied as a solvent for resins.

    Solubility
    Soluble in water. (1 g/L).

    Notes
    Store at room temperature. Incompatible with oxidizing agents.

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit