Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Butyl Chloride (Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Boiling Range | 2°C max. |
Melting Point | -123°C |
Color | Colorless |
Density | 0.880g/cm³ |
Boiling Point | 77°C |
Flash Point | −12°C |
Packaging | Glass Bottle |
Linear Formula | CH3(CH2)3Cl |
Quantity | 1 L |
UN Number | 1127 |
Show More |
Safety and Handling
Hazard Statement | DANGER! |
Precautionary Statement | Emergency Overview Flammable liquid and vapor. The toxicological properties have not been fully investigated. Wear personal protective equipment. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools. Use explosion-proof equipment. To avoid ignition of vapors by static electricity discharge, all metal parts of the equipment must b. Take precautionary measures against static discharges. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. NFPA Health:1 Flammability:3 Instability:0 |
DOTInformation | DOT Class 3, : Flammable Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction