Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Butyl chloroformate, 98%, Alfa Aesar™

Click to view available options
Quantity:
100g
250g
592-34-7
C5H9ClO2
136.575
MFCD00000650
NRDQFWXVTPZZAZ-UHFFFAOYSA-N
butyl chloroformate,n-butyl chloroformate,butylchloroformate,carbonochloridic acid, butyl ester,chloroformic acid butyl ester,butyl chlorocarbonate,n-butyl chlorocarbonate,formic acid, chloro-, butyl ester,butyl-chloroformate,butylchloro-formate
61140
butyl carbonochloridate
CCCCOC(=O)Cl
592-34-7
C5H9ClO2
136.575
MFCD00000650
NRDQFWXVTPZZAZ-UHFFFAOYSA-N
butyl chloroformate,n-butyl chloroformate,butylchloroformate,carbonochloridic acid, butyl ester,chloroformic acid butyl ester,butyl chlorocarbonate,n-butyl chlorocarbonate,formic acid, chloro-, butyl ester,butyl-chloroformate,butylchloro-formate
61140
butyl carbonochloridate
CCCCOC(=O)Cl
Odor Acrid
Quantity 250g
Boiling Point 141°C to 143°C
Flash Point 39°C (102°F)
Melting Point <-70°C
UN Number UN2743
Beilstein 605635
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material n-Butyl chloroformate

GHS H Statement
H330-H314-H318-H226
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

GHS P Statement
P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: N-BUTYL CHLOROFORMATE

TSCA : TSCA

Recommended Storage : 2° to 8°C