Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Butyl heptafluorobutyrate, 97%

Click to view available options
Quantity:
10g
50g
1559-07-5
C8H9F7O2
270.147
MFCD00039240
YDXXJZKOOOJVEE-UHFFFAOYSA-N
n-butyl heptafluorobutyrate,butyric acid, heptafluoro-, butyl ester,butyl heptafluorobutanoate,heptafluorobutyric acid, n-butyl ester,n-butylheptafluorobutyrate,butyl perfluorobutanoat,butyl perfluorobutyrate,butylheptafluorobutyrate,butyl heptafluorobutyrate,n-butyl perfluoro-n-butyrate
137079
butyl 2,2,3,3,4,4,4-heptafluorobutanoate
CCCCOC(=O)C(C(C(F)(F)F)(F)F)(F)F
1559-07-5
C8H9F7O2
270.147
MFCD00039240
YDXXJZKOOOJVEE-UHFFFAOYSA-N
n-butyl heptafluorobutyrate,butyric acid, heptafluoro-, butyl ester,butyl heptafluorobutanoate,heptafluorobutyric acid, n-butyl ester,n-butylheptafluorobutyrate,butyl perfluorobutanoat,butyl perfluorobutyrate,butylheptafluorobutyrate,butyl heptafluorobutyrate,n-butyl perfluoro-n-butyrate
137079
butyl 2,2,3,3,4,4,4-heptafluorobutanoate
CCCCOC(=O)C(C(C(F)(F)F)(F)F)(F)F
Quantity 10g
Boiling Point 131°C to 133°C
Linear Formula CF3CF2CF2CO2(CH2)3CH3
Beilstein 1798327
Formula Weight 270.15
Percent Purity 97%
Chemical Name or Material n-Butyl heptafluorobutyrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning