Learn More
n-Butyl phosphate, mixture of mono-n-butyl and di-n-butyl, Thermo Scientific Chemicals
Description
n-Butyl phosphate is a solvent and plasticizer for cellulose esters such as nitrocellulose and cellulose acetate. It forms stable hydrophobic complexes with some metals, as an extractant. It is also used in industry are as a component of aircraft hydraulic fluid and as a solvent for extraction and purification of rare earth metals from their ores.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -60°C |
Color | Colorless |
Density | 1.123 g/mL |
Boiling Point | >124°C (decomposition) |
Flash Point | 110°C (230°F) |
Refractive Index | 1.429 |
Quantity | 250 g |
UN Number | UN1718 |
Solubility Information | Soluble in ethanol,diethyl ether. Insoluble in water. |
Physical Form | Liquid |
Show More |
Safety and Handling
Hazard Category | H314-H335-H351 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BUTYL ACID PHOSPHATE |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.