Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyl vinyl ether, 98%, ACROS Organics™

Click to view available options
Quantity:
50mL
500mL
1L
Packaging:
Glass bottle
111-34-2
C6H12O
100.161
MFCD00009454
UZKWTJUDCOPSNM-UHFFFAOYSA-N
n-butyl vinyl ether,butyl vinyl ether,vinyl butyl ether,butane, 1-ethenyloxy,butoxyethylene,1-ethenyloxy butane,butoxyethene,ether, butyl vinyl,vinyl n-butyl ether,ethenyl n-butyl ether
8108
1-ethenoxybutane
CCCCOC=C
111-34-2
C6H12O
100.161
MFCD00009454
UZKWTJUDCOPSNM-UHFFFAOYSA-N
n-butyl vinyl ether,butyl vinyl ether,vinyl butyl ether,butane, 1-ethenyloxy,butoxyethylene,1-ethenyloxy butane,butoxyethene,ether, butyl vinyl,vinyl n-butyl ether,ethenyl n-butyl ether
8108
1-ethenoxybutane
CCCCOC=C
Packaging Glass bottle
Refractive Index 1.4005 to 1.4025 (20°C, 589nm)
Quantity 50mL
Stabilizer max. 1000ppm KOH
Infrared Spectrum Authentic
Color Undesignated
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)3OCH=CH2
UN Number 2352
Formula Weight 100.16
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material n-Butyl vinyl ether, Stabilized
Show More Show Less

GHS H Statement:
Extremely flammable liquid and vapour.
Causes skin irritation.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

GHS Signal Word: Danger