Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Butylbenzene, 99%

Manufacturer:  Alfa Aesar™ A1091030

Quantity 250g

 View more versions of this product

Catalog No. AAA1091030


  Check Availability
Add to cart

Specifications

Description

Applications
n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.

Solubility
Sparingly soluble in water (0.012g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Storage temperature 2 - 8°C. Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

104-51-8
134.22
UN2709
14,1549
n-Butylbenzene
C10H14
MFCD00009463
1903395
1-Phenylbutane

Specifications

104-51-8
134.22
MFCD00009463
181° to 183°C
250g
14,1549
1-Phenylbutane
99%
C10H14
−88°C
UN2709
1903395
59°C (138°F)
Aromatic
n-Butylbenzene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL BENZENES

RTECSNumber : CY9070000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit