Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Butylbenzene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.
Solubility
Sparingly soluble in water (0.012g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Storage temperature 2 - 8°C. Incompatible with strong oxidizing agents.
n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.
Solubility
Sparingly soluble in water (0.012g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Storage temperature 2 - 8°C. Incompatible with strong oxidizing agents.
Specifications
Melting Point | -88°C |
Density | 0.86 |
Boiling Point | 181°C to 183°C |
Flash Point | 59°C (138°F) |
Odor | Aromatic |
Refractive Index | 1.489 |
Quantity | 50 g |
UN Number | UN2709 |
Beilstein | 1903395 |
Merck Index | 14,1549 |
Show More |
Safety and Handling
Hazard Category | H226 |
Hazard Statement | GHS H Statement H226 Flammable liquid and vapor. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL BENZENES |
EINECSNumber | 203-209-7 |
RTECSNumber | CY9070000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction