Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butylboronic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
4426-47-5
C4H11BO2
101.94
MFCD00002106
QPKFVRWIISEVCW-UHFFFAOYSA-N
n-butylboronic acid,1-butylboronic acid,1-butaneboronic acid,n-butaneboronic acid,boronic acid, butyl,1-butane boronic acid,butylboric acid,butyl boronic acid,chembl31962,1-butyldihydroxyborane
20479
butylboronic acid
B(CCCC)(O)O
4426-47-5
C4H11BO2
101.94
MFCD00002106
QPKFVRWIISEVCW-UHFFFAOYSA-N
n-butylboronic acid,1-butylboronic acid,1-butaneboronic acid,n-butaneboronic acid,boronic acid, butyl,1-butane boronic acid,butylboric acid,butyl boronic acid,chembl31962,1-butyldihydroxyborane
20479
butylboronic acid
B(CCCC)(O)O
Acylation reagent
Melting Point 94.0°C to 96.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)3B(OH)2
Solubility Information Solubility in water:+/- 2.5% in water
Formula Weight 101.94
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material n-Butylboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-607-7