Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Butyldimethylchlorosilane, 96%

Click to view available options
Quantity:
5g
25g
100g
1000-50-6
C6H15ClSi
150.721
MFCD00015727
MXOSTENCGSDMRE-UHFFFAOYSA-N
butylchlorodimethylsilane,butyldimethylchlorosilane,n-butyldimethylchlorosilane,butyldimethylsilyl chloride,1-butyldimethylchlorosilane,silane, butylchlorodimethyl,butyl chloro dimethylsilane,n-butyl-dimethylchlorosilane,butyl-chloro-dimethyl-silane,n-butyldimethylsilyl chloride
66079
butyl-chloro-dimethylsilane
CCCC[Si](C)(C)Cl
1000-50-6
C6H15ClSi
150.721
MFCD00015727
MXOSTENCGSDMRE-UHFFFAOYSA-N
butylchlorodimethylsilane,butyldimethylchlorosilane,n-butyldimethylchlorosilane,butyldimethylsilyl chloride,1-butyldimethylchlorosilane,silane, butylchlorodimethyl,butyl chloro dimethylsilane,n-butyl-dimethylchlorosilane,butyl-chloro-dimethyl-silane,n-butyldimethylsilyl chloride
66079
butyl-chloro-dimethylsilane
CCCC[Si](C)(C)Cl
UN Number UN2920
Beilstein 1732573
Formula Weight 150.73
Percent Purity 96%
Boiling Point 138°C
Quantity 25g
Flash Point 27°C (80°F)
Chemical Name or Material n-Butyldimethylchlorosilane

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

TSCA : TSCA