n-Butyllithium, 2.2M in hexane, packaged under Argon in resealable ChemSeal™ bottles, Alfa Aesar™

Manufacturer: Alfa Aesar 41248JP

Packaging Packaged under Argon in resealable ChemSeal™ bottles
Quantity 0.05 mol

 View more versions of this product

Catalog No. AA41248JP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-72-8
    Molecular Formula C4H9Li
    Linear Formula CH3(CH2)3Li
    Formula Weight 64.06
    MDL Number MFCD00009414
    UN Number UN3394
    Beilstein 1209227
    Chemical Name or Material n-Butyllithium

    Specifications

    Formula Weight 64.06
    Quantity 0.05 mol
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H225-H250-H260-H252-H304-H361-H373-H314-H336;
    Highly flammable liquid and vapour.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities; may catch fire.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point −21°C (−7°F)
    Odor Petroleum-like
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
    Concentration 2.2M in hexane
    TSCA TSCA
    Molecular Formula C4H9Li
    Linear Formula CH3(CH2)3Li
    MDL Number MFCD00009414
    CAS 109-72-8
    UN Number UN3394
    Beilstein 1209227
    Health Hazard 1 Danger
    Color Colorless to Pale Yellow
    Packaging Packaged under Argon in resealable ChemSeal™ bottles
    Sensitivity Air and Moisture Sensitive
    Chemical Name or Material n-Butyllithium

    GHS H Statement
    H225-H250-H260-H252-H304-H361-H373-H314-H336;
    Highly flammable liquid and vapour.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities; may catch fire.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit