Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyllithium, 2.2M solution in Cyclohex./Hexane, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
109-72-8,110-82-7,110-54-3
C4H9Li
64.056
MFCD00009414
DLEDOFVPSDKWEF-UHFFFAOYSA-N
n-butyllithium,lithium, butyl,butyllithium,butyl lithium,n-butyl lithium,lithium butane,n-buli,buli,libu,unii-09w9a6b8zc
61028
lithium;butane
[Li+].CCC[CH2-]
109-72-8,110-82-7,110-54-3
C4H9Li
64.056
MFCD00009414
DLEDOFVPSDKWEF-UHFFFAOYSA-N
n-butyllithium,lithium, butyl,butyllithium,butyl lithium,n-butyl lithium,lithium butane,n-buli,buli,libu,unii-09w9a6b8zc
61028
lithium;butane
[Li+].CCC[CH2-]
Additional Information product may develop some turbidity or precipitate
Linear Formula CH3(CH2)3Li
Density 0.7500g/mL
Formula Weight 64.06
Physical Form Solution, magnesium salts may crystallize below 20°C
Percent Purity 19.5 to 20.5% (as BuLi) (Active base)
Free Alkali 1 % max. (as BuLi)
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: vigorous reaction (forming lioh and butane).
Specific Gravity 0.75
Color Brown to Gray
Quantity 100mL
Flash Point −18°C
Chemical Name or Material n-Butyllithium, 2.2M Solution in Cyclohexane
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Suspected of damaging fertility.
Catches fire spontaneously if exposed to air.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger