Learn More
n-Butyllithium, 2.2M solution in Cyclohex./Hexane, AcroSeal™, ACROS Organics™
Specifications
Additional Information | product may develop some turbidity or precipitate |
Linear Formula | CH3(CH2)3Li |
Density | 0.7500g/mL |
Formula Weight | 64.06 |
Physical Form | Solution, magnesium salts may crystallize below 20°C |
Percent Purity | 19.5 to 20.5% (as BuLi) (Active base) |
Free Alkali | 1 % max. (as BuLi) |
Packaging | AcroSeal™ Glass Bottle |
Solubility | Solubility in water: vigorous reaction (forming lioh and butane). |
Specific Gravity | 0.75 |
Show More |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Suspected of damaging fertility.
Catches fire spontaneously if exposed to air.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger