Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Butyllithium, 2.5M in hexanes, packaged in resealable septum cap bottle

Click to view available options
Quantity:
100mL
800mL
109-72-8
C4H9Li
64.056
MFCD00009414
DLEDOFVPSDKWEF-UHFFFAOYSA-N
n-butyllithium,lithium, butyl,butyllithium,butyl lithium,n-butyl lithium,lithium butane,n-buli,buli,libu,unii-09w9a6b8zc
61028
lithium;butane
[Li+].CCC[CH2-]
109-72-8
C4H9Li
64.056
MFCD00009414
DLEDOFVPSDKWEF-UHFFFAOYSA-N
n-butyllithium,lithium, butyl,butyllithium,butyl lithium,n-butyl lithium,lithium butane,n-buli,buli,libu,unii-09w9a6b8zc
61028
lithium;butane
[Li+].CCC[CH2-]
  • Packaged under Argon
Concentration 2.5M in hexanes
Quantity 100mL
Density 0.693g/mL
Color Yellow
Packaging ChemSeal™ bottles
Sensitivity Air & Moisture Sensitive
Linear Formula CH3(CH2)3Li
UN Number UN3394
Beilstein 1209227
Formula Weight 64.06
Physical Form Liquid
Chemical Name or Material n-Butyllithium
Show More Show Less

GHS H Statement
H225-H250-H260-H252-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Self-heating in large quantities; may catch fire
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packaging Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE

TSCA : TSCA

storageNote1 :

  • Packaged under Argon