Learn More
Alfa Aesar™ n-Butylmagnesium chloride, 1.5-2.5M in THF
Manufacturer:  Alfa Aesar™ 46117JQ
Quantity 0.1 mole
 View more versions of this product
Catalog No. AA46117JQ
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
693-04-9
116.87
UN3399
n-Butylmagnesium chloride
CH3(CH2)3MgCl
MFCD00000475
3587228
Specifications
693-04-9
Liquid
116.87
MFCD00000475
3587228
0.1 mole
n-Butylmagnesium chloride
Air & Moisture Sensitive
CH3(CH2)3MgCl
1.5 to 2.5M in THF
UN3399
Brown
Ether-like
Safety and Handling
SDS & Safety Documents
GHS H Statement
H224-H261-H314-H318
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : TSCA