Learn More
Alfa Aesar™ n-Butylmagnesium chloride, 1.5-2.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Grignard reagent
Manufacturer:  Alfa Aesar™ 41676AK
Quantity 250mL
 View more versions of this product
Catalog No. AA41676AK
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
693-04-9
116.87
UN3399
n-Butylmagnesium chloride
CH3(CH2)3MgCl
MFCD00000475
3587228
Specifications
693-04-9
Air & Moisture Sensitive
CH3(CH2)3MgCl
1.5 to 2.5M in THF
UN3399
Brown
n-Butylmagnesium chloride
ChemSeal™ bottles
Liquid
116.87
MFCD00000475
3587228
250mL
Safety and Handling
SDS & Safety Documents
GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
GHS P Statement
P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : TSCA
storageNote1 :
  • Packaged under Argon