Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butylmagnesium chloride, 1.5-2.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Catalog Number AA41676AD
Click to view available options
Quantity:
50 mL
250 mL
693-04-9
C4H9ClMg
116.87
MFCD00000475
UUNGXYVLXYGZKH-UHFFFAOYSA-M
butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852
101837
magnesium;butane;chloride
CCCC[Mg]Cl
693-04-9
C4H9ClMg
116.87
MFCD00000475
UUNGXYVLXYGZKH-UHFFFAOYSA-M
butylmagnesium chloride, magnesium, butylchloro, nbumgcl, bumgcl, butylmagnesiumchloride, n-butylmagnesium chloride, n-butyl-magnesium chloride, butylmagnesium chloride solution, butylmagnesium chloride solution, 2.0 m in diethyl ether, unii-4j83fgw852
101837
magnesium;butane;chloride
CCCC[Mg]Cl

Grignard reagent

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Color Brown
Packaging ChemSeal™ bottles
Linear Formula CH3(CH2)3MgCl
Quantity 50 mL
UN Number UN3399
Beilstein 3587228
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 116.87
Concentration or Composition (by Analyte or Components) 1.5 to 2.5M in THF
Physical Form Liquid
Chemical Name or Material n-Butylmagnesium chloride
Show More Show Less
Hazard Category H225-H261-H302-H314-H335-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
EINECSNumber 211-739-5
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Nitrogen
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit