Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butylmagnesium chloride, 20 wt% solution in THF/Toluene, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
693-04-9,109-99-9,108-88-3
C4H9ClMg
116.87
QUXHCILOWRXCEO-UHFFFAOYSA-M
butylmagnesium chloride,magnesium, butylchloro,nbumgcl,bumgcl,butylmagnesiumchloride,n-butylmagnesium chloride,n-butyl-magnesium chloride,butylmagnesium chloride solution,butylmagnesium chloride solution, 2.0 m in diethyl ether,unii-4j83fgw852
101837
magnesium;butane;chloride
CCC[CH2-].[Mg+2].[Cl-]
693-04-9,109-99-9,108-88-3
C4H9ClMg
116.87
QUXHCILOWRXCEO-UHFFFAOYSA-M
butylmagnesium chloride,magnesium, butylchloro,nbumgcl,bumgcl,butylmagnesiumchloride,n-butylmagnesium chloride,n-butyl-magnesium chloride,butylmagnesium chloride solution,butylmagnesium chloride solution, 2.0 m in diethyl ether,unii-4j83fgw852
101837
magnesium;butane;chloride
CCC[CH2-].[Mg+2].[Cl-]
CAS Min % 19.0,78.0,78.0
CAS Max % 22.0,81.0,81.0
Linear Formula CH3(CH2)3MgCl
Formula Weight 116.87
Physical Form Solution
Percent Purity 19 to 22% (as BUMC)
Density 1.0000g/mL
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1
Storage Information Product may develop some turbidity or precipitate
Color Brown to Brown-Green
Quantity 800mL
Flash Point −21°C
Chemical Name or Material n-Butylmagnesium chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

EINECSNumber : 211-739-5