Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-(Butylthio)acetonitrile, 95%

Click to view available options
Quantity:
5g
25g
71037-08-6
C6H11NS
129.221
MFCD00041458
LRMUBLYDYGZCDR-UHFFFAOYSA-N
n-butylthio acetonitrile,2-butylthio acetonitrile,2-butylsulfanyl acetonitrile,acetonitrile,2-butylthio,butylthio acetonitrile
7567852
2-butylsulfanylacetonitrile
CCCCSCC#N
71037-08-6
C6H11NS
129.221
MFCD00041458
LRMUBLYDYGZCDR-UHFFFAOYSA-N
n-butylthio acetonitrile,2-butylthio acetonitrile,2-butylsulfanyl acetonitrile,acetonitrile,2-butylthio,butylthio acetonitrile
7567852
2-butylsulfanylacetonitrile
CCCCSCC#N
Boiling Point 134°C to 135°C (40mmHg)
Quantity 5g
Linear Formula CH3(CH2)3SCH2CN
UN Number UN3276
Beilstein 4174357
Formula Weight 129.23
Percent Purity 95%
Chemical Name or Material n-(Butylthio)acetonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.