Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Butyltin trichloride, 96%, Thermo Scientific Chemicals

Catalog Number p-7080283
Click to view available options
Quantity:
25 g
100 g
1118-46-3
C4H9Cl3Sn
282.176
MFCD00000515
YMLFYGFCXGNERH-UHFFFAOYSA-K
butyltin trichloride, stannane, butyltrichloro, monobutyltin trichloride, butyltrichlorotin, trichlorobutyltin, butyl trichloro stannane, n-butyltin trichloride, trichlorobutylstannane, stannane, trichlorobutyl, monotributyltin trichloride
14236
butyl(trichloro)stannane
CCCC[Sn](Cl)(Cl)Cl
1118-46-3
C4H9Cl3Sn
282.176
MFCD00000515
YMLFYGFCXGNERH-UHFFFAOYSA-K
butyltin trichloride, stannane, butyltrichloro, monobutyltin trichloride, butyltrichlorotin, trichlorobutyltin, butyl trichloro stannane, n-butyltin trichloride, trichlorobutylstannane, stannane, trichlorobutyl, monotributyltin trichloride
14236
butyl(trichloro)stannane
CCCC[Sn](Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -63°C
Density 1.693 g/mL
Boiling Point 98°C (10 mmHg)
Flash Point 81°C (178°F)
Odor Acrid
Refractive Index 1.5244
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 282.17
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material n-Butyltin trichloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H372-H314-H318-H312-H227
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 214-263-6
RTECSNumber WH6780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit