Alfa Aesar™ n-Butyltrichlorosilane, 97+%

Manufacturer: Alfa Aesar™ A1125606

Quantity 5g

 View more versions of this product

Catalog No. AAA1125606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7521-80-4
    Molecular Formula C4H9Cl3Si
    Linear Formula CH3(CH2)3SiCl3
    Formula Weight 191.56
    MDL Number MFCD00013605
    UN Number UN1747
    Beilstein 1736032
    Chemical Name or Material n-Butyltrichlorosilane

    Specifications

    Formula Weight 191.56
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.
    Odor Acrid
    Linear Formula CH3(CH2)3SiCl3
    Boiling Point 142° to 143°C
    CAS 7521-80-4
    UN Number UN1747
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BUTYLTRICHLOROSILANE
    RTECS Number VV2080000
    Molecular Formula C4H9Cl3Si
    Beilstein 1736032
    Health Hazard 1 Danger
    TSCA TSCA
    MDL Number MFCD00013605
    Flash Point 45°C (113°F)
    Chemical Name or Material n-Butyltrichlorosilane
    Assay ≥97%

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit