Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cadherin Mouse anti-Human, Mouse, PE-Cyanine7, Clone: 8C11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254523PECY7

Catalog No. N254523PEC7


Add to cart

Description

Description

N-Cadherin Monoclonal antibody specifically detects N-Cadherin in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

N-Cadherin
8C11
Flow Cytometry
Conjugated, Purified
CDH2
Recombinant human N-cadherin extracellular domain (exact sequence is proprietary)
Protein A or G purified
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary
1000.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit