Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cadherin Rabbit anti-Human, Clone: 14, Novus Biologicals™

Rabbit Monoclonal Antibody

$321.00 - $493.00

Specifications

Antigen N-Cadherin
Clone 14
Dilution ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:1000
Applications ELISA, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB020987
View Documents
Novus Biologicals
NBP289753100UL
100 μg
Each for $493.00
Add to cart
 
NB020988
View Documents
Novus Biologicals
NBP28975350UL
50 μg
Each for $321.00
Add to cart
 
Description

Description

N-Cadherin Monoclonal specifically detects N-Cadherin in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

N-Cadherin
ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:1000
Monoclonal
Purified
RUO
Human
cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin
Recombinant Human N-Cadherin protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
14
ELISA, Immunofluorescence
Unconjugated
Rabbit
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction
0.2 um filtered solution in PBS
1000
IgG
Protein A purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit