Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-L-aspartic Acid 99.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
1152-61-0
C12H13NO6
267.237
MFCD00002719
XYXYXSKSTZAEJW-VIFPVBQESA-N
z-asp-oh, n-carbobenzyloxy-l-aspartic acid, cbz-asp-oh, n-carbobenzoxy-l-aspartic acid, n-cbz-l-aspartic acid, z-l-aspartic acid, carbobenzoxy-l-aspartic acid, n-z-l-aspartic acid, n-benzyloxycarbonylaspartic acid, z-l-asp-oh
2723942
(2S)-2-(phenylmethoxycarbonylamino)butanedioic acid
C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O
1152-61-0
C12H13NO6
267.237
MFCD00002719
XYXYXSKSTZAEJW-VIFPVBQESA-N
z-asp-oh, n-carbobenzyloxy-l-aspartic acid, cbz-asp-oh, n-carbobenzoxy-l-aspartic acid, n-cbz-l-aspartic acid, z-l-aspartic acid, carbobenzoxy-l-aspartic acid, n-z-l-aspartic acid, n-benzyloxycarbonylaspartic acid, z-l-asp-oh
2723942
(2S)-2-(phenylmethoxycarbonylamino)butanedioic acid
C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O
Color White
Melting Point 116°C
Quantity 25g
Formula Weight 267.24
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (T)
Chemical Name or Material N-Carbobenzoxy-L-aspartic Acid

EINECSNumber : (9)-1549

TSCA : Yes