Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Chloroacetylisonipecotic acid, 98%

Click to view available options
Quantity:
1g
5g
318280-69-2
C8H12ClNO3
205.638
MFCD02093986
PEQVHRULPWCLOJ-UHFFFAOYSA-N
n-chloroacetylisonipecotic acid,1-2-chloroacetyl piperidine-4-carboxylic acid,1-2-chloroacetyl-4-piperidinecarboxylic acid,acmc-20ak5i,n-chloroacetyl-dl-isonipecotic acid,1-chloroacetylpiperidine-4-carboxylic acid,1-chloroacetyl-4-piperidinecarboxylic acid,1-chloroacetyl piperidine-4-carboxylic acid,4-piperidinecarboxylic acid, 1-chloroacetyl,4-piperidinecarboxylic acid, 1-chloroacetyl-9ci
1991140
1-(2-chloroacetyl)piperidine-4-carboxylic acid
C1CN(CCC1C(=O)O)C(=O)CCl
318280-69-2
C8H12ClNO3
205.638
MFCD02093986
PEQVHRULPWCLOJ-UHFFFAOYSA-N
n-chloroacetylisonipecotic acid,1-2-chloroacetyl piperidine-4-carboxylic acid,1-2-chloroacetyl-4-piperidinecarboxylic acid,acmc-20ak5i,n-chloroacetyl-dl-isonipecotic acid,1-chloroacetylpiperidine-4-carboxylic acid,1-chloroacetyl-4-piperidinecarboxylic acid,1-chloroacetyl piperidine-4-carboxylic acid,4-piperidinecarboxylic acid, 1-chloroacetyl,4-piperidinecarboxylic acid, 1-chloroacetyl-9ci
1991140
1-(2-chloroacetyl)piperidine-4-carboxylic acid
C1CN(CCC1C(=O)O)C(=O)CCl
Melting Point 109°C to 111°C
Quantity 1g
Formula Weight 205.64
Percent Purity 98%
Chemical Name or Material N-Chloroacetylisonipecotic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures