Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Chlorosuccinimide, 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
128-09-6
C4H4ClNO2
133.53
JRNVZBWKYDBUCA-UHFFFAOYSA-N
n-chlorosuccinimide,succinchlorimide,chlorosuccinimide,succinochlorimide,succinic n-chloroimide,2,5-pyrrolidinedione, 1-chloro,1-chloro-2,5-pyrrolidinedione,succinimide, n-chloro,ncs,caswell no. 807
31398
CHEBI:53203
1-chloropyrrolidine-2,5-dione
C1CC(=O)N(C1=O)Cl
128-09-6
C4H4ClNO2
133.53
JRNVZBWKYDBUCA-UHFFFAOYSA-N
n-chlorosuccinimide,succinchlorimide,chlorosuccinimide,succinochlorimide,succinic n-chloroimide,2,5-pyrrolidinedione, 1-chloro,1-chloro-2,5-pyrrolidinedione,succinimide, n-chloro,ncs,caswell no. 807
31398
CHEBI:53203
1-chloropyrrolidine-2,5-dione
C1CC(=O)N(C1=O)Cl
C-X bond formation reagent
Loss on Drying 4% (2g, 110°C, 2 hrs) max.
Quantity 100g
Flash Point >110°C
Melting Point 144.0°C to 150.0°C
Color White
Packaging Plastic bottle
Infrared Spectrum Authentic
Assay Percent Range 97%
Beilstein 21,38
Fieser 01,139; 02,69; 05,127; 06,115; 07,65; 08,103; 09,111; 10,94; 12,121; 13,62; 14,87; 15,86; 16,84
Merck Index 15,2167
Solubility Information Solubility in water: 14g/L (20°C). Other solubilities: sparingly soluble in chloroform,ether and ccl4,1g/150mL alcohol- 1g/50mL benzene,soluble in acetone
Formula Weight 133.53
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material N-Chlorosuccinimide
Show More Show Less