Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Cycloheptylmethylamine, 97%

Click to view available options
Quantity:
1g
5g
42870-65-5
C8H17N
127.231
MFCD08271751
LRXSDHDEISIWQB-UHFFFAOYSA-N
n-cycloheptyl-n-methylamine,hexamethylenemethylamine,cycloheptanemethanamine,n-methylcycloheptylamine,cycloheptyl methyl amine,cycloheptyl-methyl-amine,n-methyl-n-cycloheptylamine,cycloheptanamine, n-methyl
276275
N-methylcycloheptanamine
CNC1CCCCCC1
42870-65-5
C8H17N
127.231
MFCD08271751
LRXSDHDEISIWQB-UHFFFAOYSA-N
n-cycloheptyl-n-methylamine,hexamethylenemethylamine,cycloheptanemethanamine,n-methylcycloheptylamine,cycloheptyl methyl amine,cycloheptyl-methyl-amine,n-methyl-n-cycloheptylamine,cycloheptanamine, n-methyl
276275
N-methylcycloheptanamine
CNC1CCCCCC1
Quantity 1g
Beilstein 2715358
Formula Weight 127.14
Physical Form Solid
Percent Purity 97%
Chemical Name or Material N-Cycloheptylmethylamine

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger