Alfa Aesar™ N-Cyclohexylaniline, 98%

Manufacturer: Alfa Aesar™ A1240909

Quantity 10g

 View more versions of this product

Catalog No. AAA1240909

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1821-36-9
    Molecular Formula C12H17N
    Formula Weight 175.28
    MDL Number MFCD00014284
    Beilstein 2209864
    Synonym N-Phenylcyclohexylamine
    Chemical Name or Material N-Cyclohexylaniline

    Specifications

    Formula Weight 175.28
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C12H17N
    Melting Point 14° to 15°C
    Boiling Point 279°C
    Quantity 10g
    Flash Point 136°C (277°F)
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    MDL Number MFCD00014284
    CAS 1821-36-9
    Beilstein 2209864
    Synonym N-Phenylcyclohexylamine
    Health Hazard 1 Danger
    Chemical Name or Material N-Cyclohexylaniline
    Assay 98%

    Description

    Applications
    N-Cyclohexylaniline is used as reagent in palladium-catalyzed amination of aromatic bromides. It is also used to produce N-cyclohexyl-N-phenyl-urea at ambient temperature.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit