Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Cyclohexylmaleimide, 97%

Click to view available options
Quantity:
1g
1631-25-0
C10H13NO2
179.219
MFCD00043904
BQTPKSBXMONSJI-UHFFFAOYSA-N
n-cyclohexylmaleimide,1-cyclohexyl-1h-pyrrole-2,5-dione,maleimide, n-cyclohexyl,1h-pyrrole-2,5-dione, 1-cyclohexyl,n-cyclohexyl maleimide,1-cyclohexylazoline-2,5-dione,acmc-1btr9,1h-pyrrole-2, 1-cyclohexyl,1-cyclohexyl-pyrrole-2,5-dione
74203
1-cyclohexylpyrrole-2,5-dione
C1CCC(CC1)N2C(=O)C=CC2=O
1631-25-0
C10H13NO2
179.219
MFCD00043904
BQTPKSBXMONSJI-UHFFFAOYSA-N
n-cyclohexylmaleimide,1-cyclohexyl-1h-pyrrole-2,5-dione,maleimide, n-cyclohexyl,1h-pyrrole-2,5-dione, 1-cyclohexyl,n-cyclohexyl maleimide,1-cyclohexylazoline-2,5-dione,acmc-1btr9,1h-pyrrole-2, 1-cyclohexyl,1-cyclohexyl-pyrrole-2,5-dione
74203
1-cyclohexylpyrrole-2,5-dione
C1CCC(CC1)N2C(=O)C=CC2=O
Quantity 1g
Melting Point 89°C to 91°C
Formula Weight 179.22
Percent Purity 97%
Chemical Name or Material N-Cyclohexylmaleimide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA