Alfa Aesar™ n-Decane, 99+%

Manufacturer: Alfa Aesar™ 3120909

Quantity 10g

 View more versions of this product

Catalog No. AA3120909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 124-18-5
    Molecular Formula C10H22
    Linear Formula CH3(CH2)8CH3
    Formula Weight 142.29
    MDL Number MFCD00008954
    UN Number UN2247
    Beilstein 1696981
    Chemical Name or Material n-Decane

    Specifications

    Formula Weight 142.29
    Assay Percent Range >99%
    Molecular Formula C10H22
    Health Hazard 3 GHS P Statement
    P210-P280-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    UN Number UN2247
    Beilstein 1696981
    Odor Petrol-like
    Health Hazard 2 GHS H Statement
    H304-H226
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.
    RTECS Number HD6550000
    Refractive Index 1.4115
    MDL Number MFCD00008954
    CAS 124-18-5
    Boiling Point 172° to 174°C
    Flash Point 46° (115°F)
    TSCA TSCA
    Physical Form Liquid
    Density 0.730g/mL
    Linear Formula CH3(CH2)8CH3
    Color Colorless
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-DECANE
    Melting Point −30°C
    Quantity 10g
    Chemical Name or Material n-Decane
    Assay ≥99%

    GHS H Statement
    H304-H226
    May be fatal if swallowed and enters airways.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit