n-Decane, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar A14732AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA14732AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 124-18-5
    Physical Form Liquid
    Assay Percent Range 99%
    Molecular Formula CH3(CH2)8CH3
    Formula Weight 142.29
    Chemical Name or Material n-Decane

    Specifications

    Formula Weight 142.29
    Quantity 500mL
    Boiling Point 172° to174°C
    Melting Point −30°C
    Flash Point 46°C
    Density 0.730g/mL
    Physical Form Liquid
    Assay Percent Range 99%
    Molecular Formula CH3(CH2)8CH3
    CAS 124-18-5
    Chemical Name or Material n-Decane

    Description

    Applications
    n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

    Solubility
    Soluble in alcohol and ethers. Miscible with ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit