Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Decanoic anhydride, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
2082-76-0
C20H38O3
326.521
MFCD00010460
HTWWKYKIBSHDPC-UHFFFAOYSA-N
decanoic anhydride,capric anhydride,n-capric anhydride,n-decanoic anhydride,decanoic acid anhydride,acmc-209few,decanoic acid, anhydride,decanoic acid,1,1'-anhydride,htwwkykibshdpc-uhfffaoysa
74960
decanoyl decanoate
CCCCCCCCCC(=O)OC(=O)CCCCCCCCC
2082-76-0
C20H38O3
326.521
MFCD00010460
HTWWKYKIBSHDPC-UHFFFAOYSA-N
decanoic anhydride,capric anhydride,n-capric anhydride,n-decanoic anhydride,decanoic acid anhydride,acmc-209few,decanoic acid, anhydride,decanoic acid,1,1'-anhydride,htwwkykibshdpc-uhfffaoysa
74960
decanoyl decanoate
CCCCCCCCCC(=O)OC(=O)CCCCCCCCC
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula O[C(O)(CH2)8CH3]
Formula Weight 326.52
Physical Form Melting Solid
Percent Purity ≥93%
Chemical Name or Material n-Decanoic anhydride, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 218-213-4