Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
15163-36-7
C15H33NO3S
307.5
WKALLSVICJPZTM-UHFFFAOYSA-N
3-decyldimethylammonio propane-1-sulfonate,n-decyl-n,n-dimethyl-3-ammonio-1-propanesulfonate,sulfobetaine 10,caprylyl sulfobetaine,3-decyldimethylammonio propanesulfonate,zwittergent 3-10 detergent,capryl sultaine,zwittergent 310,3-decyl dimethyl ammonio propane-1-sulfonate,3-decyldimethylamino propanesulfonic acid
161111
3-[decyl(dimethyl)azaniumyl]propane-1-sulfonate
CCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]
15163-36-7
C15H33NO3S
307.5
WKALLSVICJPZTM-UHFFFAOYSA-N
3-decyldimethylammonio propane-1-sulfonate,n-decyl-n,n-dimethyl-3-ammonio-1-propanesulfonate,sulfobetaine 10,caprylyl sulfobetaine,3-decyldimethylammonio propanesulfonate,zwittergent 3-10 detergent,capryl sultaine,zwittergent 310,3-decyl dimethyl ammonio propane-1-sulfonate,3-decyldimethylamino propanesulfonic acid
161111
3-[decyl(dimethyl)azaniumyl]propane-1-sulfonate
CCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]
Loss on Drying 2% max.
Quantity 100g
Melting Point 229.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Solubility Information (10% in water) Clear colorless solution
Formula Weight 307.5
Percent Purity 98%
Chemical Name or Material N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulfonate

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective g

GHS Signal Word: Warning