Alfa Aesar™ n-Decyldimethylchlorosilane, 97%

Manufacturer: Alfa Aesar™ 4276909

Quantity 10g

 View more versions of this product

Catalog No. AA4276909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38051-57-9
    Molecular Formula CH3(CH2)9Si(CH3)2Cl
    Formula Weight 234.88
    MDL Number MFCD00054168
    UN Number UN1760
    Chemical Name or Material n-Decyldimethylchlorosilane

    Specifications

    Formula Weight 234.88
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 38051-57-9
    Flash Point 137°C (279°F)
    Physical Form Liquid
    Refractive Index 1.441
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
    TSCA TSCA
    Sensitivity Moisture Sensitive
    MDL Number MFCD00054168
    Molecular Formula CH3(CH2)9Si(CH3)2Cl
    UN Number UN1760
    Boiling Point 98°C (2mmHg)
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Density 0.866g/mL
    Chemical Name or Material n-Decyldimethylchlorosilane
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit