Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ n-Decyltrichlorosilane, 97%

Click to view available options
Quantity:
10g
50g
13829-21-5
C10H21Cl3Si
275.713
MFCD00013600
HLWCOIUDOLYBGD-UHFFFAOYSA-N
decyltrichlorosilane, trichloro decyl silane, n-decyltrichlorosilane, silane, trichlorodecyl, acmc-209chi, trichloro n-decyl silane, trichloro decyl silane #
83764
trichloro(decyl)silane
CCCCCCCCCC[Si](Cl)(Cl)Cl
13829-21-5
C10H21Cl3Si
275.713
MFCD00013600
HLWCOIUDOLYBGD-UHFFFAOYSA-N
decyltrichlorosilane, trichloro decyl silane, n-decyltrichlorosilane, silane, trichlorodecyl, acmc-209chi, trichloro n-decyl silane, trichloro decyl silane #
83764
trichloro(decyl)silane
CCCCCCCCCC[Si](Cl)(Cl)Cl
Linear Formula CH3(CH2)9SiCl3
UN Number UN2987
Beilstein 1749722
Formula Weight 275.72
Percent Purity 97%
Density 1.049
Refractive Index 1.452
Sensitivity Moisture sensitive
Boiling Point 133°C to 137°C (5mmHg)
Quantity 10g
Flash Point >110°C (230°F)
Chemical Name or Material n-Decyltrichlorosilane
Show More Show Less

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

EINECSNumber : 237-540-3

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen